Learn More
Niacinamide, USP, 98.5-101.5%, Spectrum™ Chemical
NI105, 98-92-0, C6H6N2O
Supplier:  Spectrum Chemical Mfg Cor NI1055KG
 View more versions of this product
Catalog No. 18604794
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Niacinamide, USP is a vitamin supplement. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
98-92-0
0.001
128-131°C
C6H6N2O
DFPAKSUCGFBDDF-UHFFFAOYSA-N
pyridine-3-carboxamide
98.5 to 101.5%
1
0.005
Poly Pail
5 kg
NC(=O)C1=CC=CN=C1
122.13
USP
Chemical Identifiers
98-92-0
122.13
pyridine-3-carboxamide
C6H6N2O
DFPAKSUCGFBDDF-UHFFFAOYSA-N
NC(=O)C1=CC=CN=C1
Product Suggestions
Product Suggestions