missing translation for 'onlineSavingsMsg'
Learn More

Niacinamide, USP, 98.5-101.5%, Spectrum™ Chemical

NI105, 98-92-0, C6H6N2O

$234.66 - $581.06

Chemical Identifiers

CAS 98-92-0
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
InChI Key DFPAKSUCGFBDDF-UHFFFAOYSA-N
IUPAC Name pyridine-3-carboxamide
SMILES NC(=O)C1=CC=CN=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18604791
View Documents
Spectrum Chemical Mfg Cor
NI105100GM
100 g Poly Bottle
Each for $234.66
Only null left
 
18604793
View Documents
Spectrum Chemical Mfg Cor
NI1051KG
1 kg Poly Pail
Each for $581.06
Only null left
 
Description

Description

Spectrum™ Chemical Niacinamide, USP is a vitamin supplement. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

98-92-0
122.13
pyridine-3-carboxamide
C6H6N2O
DFPAKSUCGFBDDF-UHFFFAOYSA-N
NC(=O)C1=CC=CN=C1

Specifications

98-92-0
0.001
128-131°C
C6H6N2O
DFPAKSUCGFBDDF-UHFFFAOYSA-N
pyridine-3-carboxamide
98.5 to 101.5%
1
0.005
Poly Bottle
100 g
NC(=O)C1=CC=CN=C1
122.13
USP
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'