missing translation for 'onlineSavingsMsg'
Learn More

NDUFB8 Monoclonal Antibody (K9N014_11C6), Invitrogen™

Mouse Monoclonal Antibody

$1693.48 - $2847.38

Specifications

Antigen NDUFB8
Clone K9N014_11C6
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MA556007
View Documents
Invitrogen™
MA556007
200 μg
Each for $2,847.38
Only null left
 
MA555627
View Documents
Invitrogen™
MA555627
50 μg
Each for $1,693.48
Only null left
 
Description

Description

Sequence of this protein is as follows: YPRTPEERAA AAKKYNMRVE DYEPYPDDGM GYGDYPKLPD RSQHERDPWY SWDQPGLRLN WGEPMHWHLD

NDUFB8 Monoclonal antibody specifically detects NDUFB8 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

NDUFB8
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide
19 kDa; 2900010I05Rik; AI987932; ASHI; CI-ASHI; complex I ASHI subunit; complex I-ASHI; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 8; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 8, 19kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 8, mitochondrial; NADH dehydrogenase 1 beta subcomplex 8; NADH:ubiquinone oxidoreductase ASHI subunit; NADH:ubiquinone oxidoreductase subunit B8; NADH-ubiquinone oxidoreductase ASHI subunit; NDUFB8
Recombinant human NDUFB8 (immunogen range is 41-110).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
NDUFB8
K9N014_11C6
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Mouse, Rat
O95169, Q9D6J5
293991, 4714, 67264
IgG1
Protein A
Antibody
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'