missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Reagent Bottles
Rigid bottles for universities and industrial labs
$19.00 - $19.00
Specifications
Capacity (Metric) | 500 mL |
---|---|
Capacity (English) | 17 oz. |
Color | Clear |
Description | Leak-proof |
Material | Polypropylene |
Description
- Made from polypropylene
- Available as wide or narrow neck
- Autoclavable
- Leak-proof
Specifications
500 mL | |
Clear | |
Polypropylene | |
1 Ea. | |
6.98 cm | |
Autoclavable | |
17.14 cm | |
6.75 in. |
17 oz. | |
Leak-proof | |
6.98 cm | |
Non-sterile | |
Yes | |
No | |
Reagent Bottle |