missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-(tert-Butoxycarbonyl)-L-arginine Hydrochloride Monohydrate 98.0+%, TCI America™
$63.38 - $167.76
Chemical Identifiers
CAS | 35897-34-8 |
---|---|
Molecular Formula | C11H23ClN4O4 |
Molecular Weight (g/mol) | 310.78 |
MDL Number | MFCD00065552 |
InChI Key | HDELGKMVZYHPPB-RZUXQYDTNA-N |
Synonym | boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, n-boc-l-arginine hydrochloride monohydrate, boc-d-arg-oh hcl h2o, boc-arg-oh hcl h2o, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, pubchem14932, pubchem14932 |
PubChem CID | 2729004 |
IUPAC Name | (2S)-2-{[(tert-butoxy)carbonyl]amino}-5-carbamimidamidopentanoic acid hydrochloride |
SMILES | Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O |
Chemical Identifiers
35897-34-8 | |
310.78 | |
HDELGKMVZYHPPB-RZUXQYDTNA-N | |
2729004 | |
Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O |
C11H23ClN4O4 | |
MFCD00065552 | |
boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, n-boc-l-arginine hydrochloride monohydrate, boc-d-arg-oh hcl h2o, boc-arg-oh hcl h2o, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, pubchem14932, pubchem14932 | |
(2S)-2-{[(tert-butoxy)carbonyl]amino}-5-carbamimidamidopentanoic acid hydrochloride |
Specifications
35897-34-8 | |
C11H23ClN4O4 | |
5 g | |
HDELGKMVZYHPPB-RZUXQYDTNA-N | |
(2S)-2-{[(tert-butoxy)carbonyl]amino}-5-carbamimidamidopentanoic acid hydrochloride | |
2729004 | |
≥98.0% (T) | |
Nalpha-(tert-Butoxycarbonyl)-L-arginine Hydrochloride Monohydrate |
White | |
MFCD00065552 | |
boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, n-boc-l-arginine hydrochloride monohydrate, boc-d-arg-oh hcl h2o, boc-arg-oh hcl h2o, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, pubchem14932, pubchem14932 | |
Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O | |
310.78 | |
310.78 | |
Crystalline Powder |
Safety and Handling
TSCA : No