missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-(tert-Butoxycarbonyl)-D-arginine Hydrochloride Monohydrate 98.0+%, TCI America™
$70.84 - $205.05
Chemical Identifiers
CAS | 114622-81-0 |
---|---|
Molecular Formula | C11H25ClN4O5 |
Molecular Weight (g/mol) | 328.794 |
MDL Number | MFCD00065554 |
InChI Key | OVXLPYFDJUFEHQ-KLXURFKVSA-N |
Synonym | boc-l-arg-oh.hcl.h2o, boc-l-arg-ohhclh2o, boc-arg-oh.hcl.h2o, boc-arg-oh.hcl.h2o, boc-arg-oh hcl h2o, boc-arg-oh hcl h2o, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, n-, pubchem14932, pubchem14932 |
PubChem CID | 2729004 |
IUPAC Name | (2S)-5-(diaminomethylideneamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate;hydrochloride |
SMILES | CC(C)(C)OC(=O)NC(CCCN=C(N)N)C(=O)O.O.Cl |
Chemical Identifiers
114622-81-0 | |
328.794 | |
OVXLPYFDJUFEHQ-KLXURFKVSA-N | |
2729004 | |
CC(C)(C)OC(=O)NC(CCCN=C(N)N)C(=O)O.O.Cl |
C11H25ClN4O5 | |
MFCD00065554 | |
boc-l-arg-oh.hcl.h2o, boc-l-arg-ohhclh2o, boc-arg-oh.hcl.h2o, boc-arg-oh.hcl.h2o, boc-arg-oh hcl h2o, boc-arg-oh hcl h2o, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, n-, pubchem14932, pubchem14932 | |
(2S)-5-(diaminomethylideneamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate;hydrochloride |
Specifications
114622-81-0 | |
C11H25ClN4O5 | |
1 g | |
OVXLPYFDJUFEHQ-KLXURFKVSA-N | |
(2S)-5-(diaminomethylideneamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate;hydrochloride | |
2729004 | |
≥98.0% (N) | |
Nalpha-(tert-Butoxycarbonyl)-D-arginine Hydrochloride Monohydrate |
White | |
MFCD00065554 | |
boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, boc-arg-oh.hcl.h2o, boc-d-arg-oh hcl h2o, boc-d-arg-oh hcl h2o, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, a-boc-d-arginine hydrochloride monohydrate, pubchem14932, pubchem14932 | |
CC(C)(C)OC(=O)NC(CCCN=C(N)N)C(=O)O.O.Cl | |
328.794 | |
310.78 | |
Crystalline Powder |
Safety and Handling
TSCA : No