Learn More
Thermo Scientific™ Nalgene™ Septum Closures: Sterile, Bulk Pack, 38-430mm
Combine elastomeric sealing of silicone with nonreactive PTFE reagent contact surfaces in these septum closures for media bottles.
$1673.26 - $1673.26
Specifications
Color | White |
---|---|
Description | Nalgene Closure for Media Bottles |
Material | HDPE |
Packaging | Bulk |
Sterility | Sterile |
Description
Combine the elastomeric sealing of silicone with nonreactive polytetrafluoroethylene reagent contact surfaces in Thermo Scientific™ Nalgene™ Septum Closures, for use with Nalgene™ Sterile PET & PETG media bottles.
Features:- Sterile bottles with molded-in graduations and neck-ring
- Non-cytotoxic, non-hemolytic and non-pyrogenic
- Display excellent gas barrier properties
- Gamma irradiated to 10-6 Sterility Assurance Level (SAL)
- Made from ADC (Animal Derived Component) Free material
- Bottles are packed without closures - order separately 342151-0384
Note:
Bottles are tested during each manufacturing run to ensure leakproof† performance. Suitable for air shipment. †The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
White | |
HDPE | |
Sterile | |
Manufactured in compliance with ISO 13485 quality requirements | |
Septum Closure |
Nalgene Closure for Media Bottles | |
Bulk | |
Silicone, PTFE | |
Sterile | |
Nalgene Sterile PET and PETG media bottles |