missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles with Closure: Bulk Pack

Ship and store liquid reagents using these high density, durable polyethylene bottles with closure. Recommended for customers who design, assemble and certify their own combination packaging.

$2275.12 - $5054.45

Specifications

Color Translucent
Description IP2 Narrow-Mouth Bottles
For Use With (Application) Storage and Shipment of Liquid Reagents
Material HDPE
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9.
View More Specs

Products 4
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
03-313-162
View Documents
Thermo Scientific™
3120990001
30 mL 20-415 mm
Case of 1000 Each for $4,016.03
Only null left
 
14-178-181
View Documents
Thermo Scientific™
3120990002
60 mL 20-415 mm
Case of 1000 Each for $5,054.45
Only null left
 
03-313-32C
View Documents
Thermo Scientific™
3120990004
125 mL 24-415 mm
Case of 500 Each for $2,857.39
Only null left
 
03-313-32D
View Documents
Thermo Scientific™
3120990008
250 mL 24-415 mm
Case of 250 Each for $2,275.12
Only null left
 
Description

Description

Ship and store liquid reagents using durable Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles with Closure. These high density polyethylene bottles are a safe, economical solution for customers who design, assemble and certify their own combination packaging.

Features:
  • HDPE bottle with PP closure, bulk pack
  • Narrow mouth
  • Complies with IATA and ICAO regulations

Ordering Information:

Bottles and closures are bulk packed in separate polybags within the same carton

Recommended for:

Storage and shipment of liquid reagents

Specifications

Specifications

Translucent
Storage and Shipment of Liquid Reagents
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9.
Narrow
IP2 Bottle
IP2 Narrow-Mouth Bottles
HDPE
Yes
Round
Polypropylene
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'