Learn More
Thermo Scientific™ Nalgene™ Large Narrow-Mouth Amber Bottles
Protect light-sensitive liquids with these amber bottles that reduce UV light transmissions and feature excellent chemical resistance to most acids, bases and alcohols.
$98.45 - $224.77
Specifications
Color Amber
Description Amber Narrow-Mouth Bottle
Material PPCO
No. per Case 6
No. per Pack 1
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
02-884-6A
View Documents
Thermo Scientific™
22040005
2 L 38-430 mm
Each for $98.45
Case of 6 Each for $590.70
Only null left
02-884-6B
View Documents
Thermo Scientific™
22040010
4 L 38-430 mm
Each for $134.48
Case of 6 Each for $806.88
Only null left
02-884-6D
View Documents
Thermo Scientific™
22040020
8 L 53B
Each for $224.77
Case of 6 Each for $1,348.62
Only null left
Description
Description
Protect light-sensitive liquids with Thermo Scientific™ Nalgene™ Large Amber Narrow-Mouth Bottles, which reduce UV light transmissions. Useful for storing chemicals and standards, as well as for collecting and storing distilled water, these bottles display excellent chemical resistance to most acids, bases and alcohols.
Features:
  • Opaque amber reduces UV light transmissions to protect light-sensitive liquids
  • Meet the U.S. Pharmacopoeia 23 requirements for light transmission
  • Excellent chemical resistance to most acids, bases and alcohols make these versatile bottles where exposure to light needs to be restricted
  • 4 and 8L sizes feature a built-in shoulder loop on larger sizes for attaching identification tag
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
  • Autoclavable/Leakproof† when used with Nalgene closures
  • Bottle and carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Note:
Completely disengage threads or remove cap before autoclaving. Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.
Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.
Specifications
Specifications
Amber
PPCO
1
Comply with U.S. Pharmacopeia XXIII requirements for maximum allowable light transmission
Autoclavable
Round
Unlined
No
Amber Narrow-Mouth Bottle
6
Non-sterile
Yes
Narrow
Narrow Mouth Bottle
Polypropylene
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.