Learn More
Thermo Scientific™ Nalgene™ Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure: Lab Pack
Effectively store high-purity chemicals with these HDPE bottles, which are extremely low in trace metal content.
$821.13 - $983.42
Specifications
Color | Natural |
---|---|
Description | Low-Particulate Narrow-Mouth Bottles |
Material | HDPE |
Sterility | Non-sterile |
Closure Color | Natural |
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
03-313-87B
|
Thermo Scientific™
3820990250 |
250 mL | 24 mm |
N/A
|
|
||||
03-313-87C
|
Thermo Scientific™
3820990500 |
500 mL | 28 mm |
N/A
|
|
||||
03-313-87D
|
Thermo Scientific™
3820991000 |
1 L | 38-430 mm |
N/A
|
|
||||
Description
Thermo Scientific™ Nalgene Certified Low Particulate Narrow-Mouth HDPE Bottles are extremely low in trace metal content, making them an excellent choice for storing high-purity chemicals. Controlled environment manufacturing minimizes particulate contamination. Narrow-mouth bottles with certified low-particulate levels are used by shippers who design and assemble combination packaging for high-purity chemicals.
Features:- Leakproof† when used with Nalgene closures
Includes:
Polypropylene closures
Recommended for:
Storage and shipping of microelectronic or biological reagents
A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database
Specifications
Natural | |
HDPE | |
Natural | |
Yes | |
Round | |
Polypropylene | |
No |
Low-Particulate Narrow-Mouth Bottles | |
Non-sterile | |
Yes | |
Narrow | |
Narrow Mouth Bottle | |
Screw Closure (Assembled) |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.