Learn More
Thermo Scientific™ Nalgene™ PC Platinum Certified Clean™ Biotainer™ Bottle
Thermo Scientific™ Nalgene™ PC Platinum Certified Clean™ Biotainer™ Bottles are certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination.
$2523.20 - $37284.57
Specifications
For Use With (Application) | Media/Buffer Preparation â, Media/Buffer Storage, Sampling, Storage, Transportation â, Validation Testing, Drug Substance, Drug Product |
---|---|
Material | Polycarbonate |
Sterility | Sterile |
Temperature Range (Metric) | -100°C to 100°C |
Closure Color | White |
Catalog Number | Mfr. No. | Capacity (Metric) | Shape | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Shape | Price | Quantity | ||||
14388043
|
Thermo Scientific™
CEN350005 |
5 mL | Round |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
14388044
|
Thermo Scientific™
CEN350020 |
20 mL | Round |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
14388040
|
Thermo Scientific™
CEN341042 |
10 L | Square |
Case of 2 Each for $2,523.20
|
|
||||
Description
Products are washed with USP Water for Injection and handled post-washing in Class 5 clean rooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.
- Reduce the risk of carry-over contamination and eliminate the cost of cleaning, sterilizing and associated validations
- Durable, translucent PC construction ensures the integrity of your sample
- Designed specifically for pharmaceutical and biological manufacturers
- Closures feature a silicone liner for leakproof assurance
- Extensive validation documentation support
- Excellent for freezing applications
Specifications
Media/Buffer Preparation â, Media/Buffer Storage, Sampling, Storage, Transportation â, Validation Testing, Drug Substance, Drug Product | |
Sterile | |
White | |
Yes | |
Narrow | |
Silicone |
Polycarbonate | |
-100°C to 100°C | |
<1/3 USP <788> (Ultralow particulate) | |
Not Autoclavable | |
Media Bottle | |
Polypropylene |