missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Undecane, 99.7%, MP Biomedicals™
$453.72 - $453.72
Chemical Identifiers
CAS | 1120-21-4 |
---|---|
Molecular Formula | C11H24 |
Molecular Weight (g/mol) | 156.31 |
MDL Number | MFCD00008959 |
InChI Key | RSJKGSCJYJTIGS-UHFFFAOYSA-N |
Synonym | n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und |
PubChem CID | 14257 |
ChEBI | CHEBI:46342 |
IUPAC Name | undecane |
SMILES | CCCCCCCCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN15823190
|
MP Biomedicals Inc
0215823190 |
500 mL |
Each for $453.72
|
|
|||||
Chemical Identifiers
1120-21-4 | |
156.31 | |
RSJKGSCJYJTIGS-UHFFFAOYSA-N | |
14257 | |
undecane |
C11H24 | |
MFCD00008959 | |
n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und | |
CHEBI:46342 | |
CCCCCCCCCCC |
Specifications
1120-21-4 | |
0.744 at 25°C (water = 1) | |
C11H24 | |
500 mL | |
RSJKGSCJYJTIGS-UHFFFAOYSA-N | |
undecane | |
14257 | |
156.3 | |
n-Undecane |
Colorless | |
0.997 | |
MFCD00008959 | |
n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und | |
CCCCCCCCCCC | |
156.31 | |
CHEBI:46342 | |
Liquid |