missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Tetradecane, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor T200725LTGL
Specifications
629-59-4 | |
Glass Bottle | |
0.99 |
1 | |
2.5 L | |
Reagent |
Chemical Identifiers
629-59-4 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.