missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl Alcohol, ACS, 99.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P1430200LTBL

 View more versions of this product

Catalog No. 18-612-048


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

71-23-8
Drum
200 L
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
0.995
0.0004 meq/g
1
C3H8O
1e-05
CCCO
60.10
ACS

Chemical Identifiers

71-23-8
60.10
propan-1-ol
C3H8O
BDERNNFJNOPAEC-UHFFFAOYSA-N
CCCO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.