missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Propyl Acetate, Reagent Grade, Reagents
N-Propyl Acetate, Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$479.40 - $2195.20
Chemical Identifiers
CAS | 109-60-4 |
---|---|
Molecular Formula | C5H10O2 |
Molecular Weight (g/mol) | 102.13 |
InChI Key | YKYONYBAUNKHLG-UHFFFAOYSA-N |
Synonym | Propyl Ethanoate, Acetic Acid Propyl Ester |
IUPAC Name | propyl acetate |
SMILES | CCCOC(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-338-133
|
Labchem Inc
C112101010E6 |
10 L |
Each for $1,508.97
|
|
|||||
01-338-132
|
Labchem Inc
C112101020E6 |
20 L |
Each for $2,195.20
|
|
|||||
01-338-131
|
Labchem Inc
C11210104C |
4 L |
Each for $479.40
|
|
|||||
Chemical Identifiers
109-60-4 | |
102.13 | |
Propyl Ethanoate, Acetic Acid Propyl Ester | |
CCCOC(C)=O |
C5H10O2 | |
YKYONYBAUNKHLG-UHFFFAOYSA-N | |
propyl acetate |
Specifications
109-60-4 | |
Colorless | |
Steel Pail | |
10 L | |
Propyl Ethanoate, Acetic Acid Propyl Ester | |
CCCOC(C)=O | |
102.13 | |
Liquid |
-93°C | |
101.5°C | |
C5H10O2 | |
0.886 | |
YKYONYBAUNKHLG-UHFFFAOYSA-N | |
propyl acetate | |
Reagent | |
N-Propyl Acetate, Reagent Grade |