missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylaniline, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor D102525LTPV
Specifications
121-69-7 | |
0.956 g/mL | |
2.5 L | |
Reagent |
1 | |
Poly-Coated Glass Bottle | |
0.99 |
Chemical Identifiers
121-69-7 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.