missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylacetamide Dimethyl Acetal (stabilized with 5-10% Methanol) 90.0+%, TCI America™
$143.54 - $432.46
Chemical Identifiers
CAS | 18871-66-4 |
---|---|
Molecular Formula | C6H15NO2 |
Molecular Weight (g/mol) | 133.191 |
MDL Number | MFCD00008476 |
InChI Key | FBZVZUSVGKOWHG-UHFFFAOYSA-N |
Synonym | n,n-dimethylacetamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylethylamine, 1,1-dimethoxyethyl dimethylamine, 1,1-dimethoxyethyl dimethyl amine, 1,1-dimethoxy-ethyl-dimethyl-amine, ethanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylacetamide dimethylacetal, dimethylacetamide dimethyl acetal, n,n-dimethylacetamide dimethyl acetal, stabilized with methanol, zlchem 332 |
PubChem CID | 87835 |
IUPAC Name | 1,1-dimethoxy-N,N-dimethylethanamine |
SMILES | CC(N(C)C)(OC)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
D165225ML
|
TCI America
D165225ML |
25 mL |
Each for $143.54
|
|
|||||
D1652100ML
|
TCI America
D1652100ML |
100 mL |
Each for $432.46
|
|
|||||
Chemical Identifiers
18871-66-4 | |
133.191 | |
FBZVZUSVGKOWHG-UHFFFAOYSA-N | |
87835 | |
CC(N(C)C)(OC)OC |
C6H15NO2 | |
MFCD00008476 | |
n,n-dimethylacetamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylethylamine, 1,1-dimethoxyethyl dimethylamine, 1,1-dimethoxyethyl dimethyl amine, 1,1-dimethoxy-ethyl-dimethyl-amine, ethanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylacetamide dimethylacetal, dimethylacetamide dimethyl acetal, n,n-dimethylacetamide dimethyl acetal, stabilized with methanol, zlchem 332 | |
1,1-dimethoxy-N,N-dimethylethanamine |
Specifications
18871-66-4 | |
C6H15NO2 | |
25 mL | |
n,n-dimethylacetamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylethylamine, 1,1-dimethoxyethyl dimethylamine, 1,1-dimethoxyethyl dimethyl amine, 1,1-dimethoxy-ethyl-dimethyl-amine, ethanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylacetamide dimethylacetal, dimethylacetamide dimethyl acetal, n,n-dimethylacetamide dimethyl acetal, stabilized with methanol, zlchem 332 | |
CC(N(C)C)(OC)OC | |
133.191 | |
133.19 | |
Liquid |
Yellow | |
MFCD00008476 | |
1993 | |
FBZVZUSVGKOWHG-UHFFFAOYSA-N | |
1,1-dimethoxy-N,N-dimethylethanamine | |
87835 | |
≥90.0% (T) | |
N,N-Dimethylacetamide Dimethyl Acetal (stabilized with 5-10% Methanol) |
Safety and Handling
TSCA : No