missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethyl-3,4,9,10-perylenetetracarboxylic Diimide 92.0+%, TCI America™

$258.90 - $795.59

Chemical Identifiers

CAS 5521-31-3
Molecular Formula C26H14N2O4
Molecular Weight (g/mol) 418.408
MDL Number MFCD00071975
InChI Key PJQYNUFEEZFYIS-UHFFFAOYSA-N
Synonym Pigment Red 179
PubChem CID 79657
SMILES CN1C(=O)C2=C3C(=CC=C4C3=C(C=C2)C5=C6C4=CC=C7C6=C(C=C5)C(=O)N(C7=O)C)C1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D44291G
View Documents
TCI America
D44291G
1 g
Each for $258.90
Only null left
 
D44295G
View Documents
TCI America
D44295G
5 g
Each for $795.59
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5521-31-3
418.408
PJQYNUFEEZFYIS-UHFFFAOYSA-N
79657
C26H14N2O4
MFCD00071975
Pigment Red 179
CN1C(=O)C2=C3C(=CC=C4C3=C(C=C2)C5=C6C4=CC=C7C6=C(C=C5)C(=O)N(C7=O)C)C1=O

Specifications

5521-31-3
MFCD00071975
Pigment Red 179
CN1C(=O)C2=C3C(=CC=C4C3=C(C=C2)C5=C6C4=CC=C7C6=C(C=C5)C(=O)N(C7=O)C)C1=O
79657
≥92.0% (N)
N,N′-Dimethyl-3,4,9,10-perylenetetracarboxylic Diimide
C26H14N2O4
1 g
PJQYNUFEEZFYIS-UHFFFAOYSA-N
418.408
418.41
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-1112&(5)-3278

RTECSNumber : CB1590000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'