missing translation for 'onlineSavingsMsg'
Learn More

N-Methylbis(trifluoroacetamide) 97.0+%, TCI America™

$113.71 - $318.75

Chemical Identifiers

CAS 685-27-8
Molecular Formula C5H3F6NO2
Molecular Weight (g/mol) 223.074
MDL Number MFCD00000412
InChI Key AWGBWLXGUPTXHF-UHFFFAOYSA-N
Synonym mbtfa, n-methyl-bis trifluoroacetamide, n-methylbis trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl acetamide, n,n-bis trifluoroacetyl methylamine, 2,2,2-trifluoro-n-methyl-n-2,2,2-trifluoroacetyl acetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl, n-methylbistrifluoroacetamide, n-methyl-bis-trifluoroacetamide
PubChem CID 69635
IUPAC Name 2,2,2-trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide
SMILES CN(C(=O)C(F)(F)F)C(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M06711ML
View Documents
TCI America
M06711ML
1 mL
Each for $113.71
Only null left
 
M06715ML
View Documents
TCI America
M06715ML
5 mL
Each for $318.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

685-27-8
223.074
AWGBWLXGUPTXHF-UHFFFAOYSA-N
69635
CN(C(=O)C(F)(F)F)C(=O)C(F)(F)F
C5H3F6NO2
MFCD00000412
mbtfa, n-methyl-bis trifluoroacetamide, n-methylbis trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl acetamide, n,n-bis trifluoroacetyl methylamine, 2,2,2-trifluoro-n-methyl-n-2,2,2-trifluoroacetyl acetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl, n-methylbistrifluoroacetamide, n-methyl-bis-trifluoroacetamide
2,2,2-trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide

Specifications

685-27-8
C5H3F6NO2
1 mL
mbtfa, n-methyl-bis trifluoroacetamide, n-methylbis trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl acetamide, n,n-bis trifluoroacetyl methylamine, 2,2,2-trifluoro-n-methyl-n-2,2,2-trifluoroacetyl acetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl, n-methylbistrifluoroacetamide, n-methyl-bis-trifluoroacetamide
CN(C(=O)C(F)(F)F)C(=O)C(F)(F)F
223.074
223.07
Liquid
Yellow
MFCD00000412
2920
AWGBWLXGUPTXHF-UHFFFAOYSA-N
2,2,2-trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide
69635
≥97.0% (GC)
N-Methylbis(trifluoroacetamide) [Trifluoroacylating Agent]
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'