missing translation for 'onlineSavingsMsg'
Learn More

N-methyl-bis(trifluoroacetamide), 98%

CAS: 685-27-8 | C5H3F6NO2 | 223.07 g/mol

$268.99 - $949.69

Chemical Identifiers

CAS 685-27-8
Molecular Formula C5H3F6NO2
Molecular Weight (g/mol) 223.07
InChI Key AWGBWLXGUPTXHF-UHFFFAOYSA-N
Synonym mbtfa, n-methyl-bis trifluoroacetamide, n-methylbis trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl acetamide, n,n-bis trifluoroacetyl methylamine, 2,2,2-trifluoro-n-methyl-n-2,2,2-trifluoroacetyl acetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl, n-methylbistrifluoroacetamide, n-methyl-bis-trifluoroacetamide
PubChem CID 69635
IUPAC Name 2,2,2-trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide
SMILES CN(C(=O)C(F)(F)F)C(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC210070050
SDS View Documents
Thermo Scientific Chemicals
210070050
5 mL Glass bottle
Each for $268.99
Only null left
 
AC210070250
SDS View Documents
Thermo Scientific Chemicals
210070250
25 mL Glass Bottle
Each for $949.69
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

685-27-8
223.07
mbtfa, n-methyl-bis trifluoroacetamide, n-methylbis trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl acetamide, n,n-bis trifluoroacetyl methylamine, 2,2,2-trifluoro-n-methyl-n-2,2,2-trifluoroacetyl acetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl, n-methylbistrifluoroacetamide, n-methyl-bis-trifluoroacetamide
2,2,2-trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide
C5H3F6NO2
AWGBWLXGUPTXHF-UHFFFAOYSA-N
69635
CN(C(=O)C(F)(F)F)C(=O)C(F)(F)F

Specifications

685-27-8
120°C to 122°C
97.5% min. (GC)
C5H3F6NO2
5 mL
mbtfa, n-methyl-bis trifluoroacetamide, n-methylbis trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl acetamide, n,n-bis trifluoroacetyl methylamine, 2,2,2-trifluoro-n-methyl-n-2,2,2-trifluoroacetyl acetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl, n-methylbistrifluoroacetamide, n-methyl-bis-trifluoroacetamide
CN(C(=O)C(F)(F)F)C(=O)C(F)(F)F
223.07
223.07
N-Methyl-bis(trifluoroacetamide)
1.5470g/mL
45°C
Glass bottle
(CF3CO)2NCH3
1.547
AWGBWLXGUPTXHF-UHFFFAOYSA-N
2,2,2-trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide
69635
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 211-680-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only