missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, PRF, B&J Brand, meets requirements of ASTM D2699 and D2700, >99.75%, Honeywell Burdick & Jackson

$8086.08 - $13284.80

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6000633
View Documents
Honeywell Inc
RS21019
19 L Steel Can
Each for $8,086.08
Only null left
 
6000634
View Documents
Honeywell-Burdick and Jackson
RS210204
204 L Steel Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

142-82-5
100.21
IMNFDUFMRHMDMM-UHFFFAOYSA-N
8900
heptane
C7H16
MFCD00009544
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CHEBI:43098
CCCCCCC

Specifications

142-82-5
0.684 g/cm3
19 L
Steel Can
MFCD00009544
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CCCCCCC
100.21
CHEBI:43098
>99.75%
n-Heptane, PRF, B&J Brand, meets requirements of ASTM D2699 and D2700, >99.75%
100.0000%
98.4°C
−4°C
C7H16
UN1206
IMNFDUFMRHMDMM-UHFFFAOYSA-N
heptane
8900
100.21
Primary Reference Fuel
Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Heptanes

SDS
missing translation for 'documents'

missing translation for 'documents'