missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ N-Ferno™ 6480 Bottoms
For lower body temperature and moisture management.
$64.52 - $64.52
Specifications
Color | Black |
---|---|
For Use With (Application) | Road Construction, Heavy Construction,Building Construction, Iron/Steel Work, Oil/Gas Refining, Drilling/Mining, Freight/Baggage, Warehousing/Distribution, Landscaping/Grounds, Shakespeare In The Park |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-910-187
|
Ergodyne™
40807 |
3X-Large |
Each for $64.52
|
|
|||||
Specifications
Black | |
Road Construction, Heavy Construction,Building Construction, Iron/Steel Work, Oil/Gas Refining, Drilling/Mining, Freight/Baggage, Warehousing/Distribution, Landscaping/Grounds, Shakespeare In The Park |