missing translation for 'onlineSavingsMsg'
Learn More

n-Butyric Acid, FCC, 99%, Spectrum™ Chemical

B2307, 107-92-6, C4H8O2

$735.94 - $6309.71

Chemical Identifiers

CAS 107-92-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002814
InChI Key FERIUCNNQQJTOY-UHFFFAOYSA-N
IUPAC Name butanoic acid
SMILES CCCC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18604766
View Documents
Spectrum Chemical Mfg Cor
B23071KGBL
1 kg Amber Glass Bottle
Each for $735.94
Only null left
 
18604765
View Documents
Spectrum Chemical Mfg Cor
B230745KGBL
45 kg Fiber Drum
Each for $6,309.71
Only null left
 
Description

Description

Spectrum™ Chemical n-Butyric Acid, FCC is used as an animal feed supplement due to the ability to reduce pathogenic bacterial colonization and is an approved food flavoring in the EU FLAVIS database (number 08.005). The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

107-92-6
88.11
FERIUCNNQQJTOY-UHFFFAOYSA-N
CCCC(O)=O
C4H8O2
MFCD00002814
butanoic acid

Specifications

107-92-6
C4H8O2
FERIUCNNQQJTOY-UHFFFAOYSA-N
butanoic acid
99%
1 kg
1.397 to 1.399
1
MFCD00002814
CCCC(O)=O
88.11
FCC
Amber Glass Bottle
0.953-0.957
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'