missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butyric Acid, 98%, Spectrum™ Chemical
$239.03 - $23175.20
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18611913
|
Spectrum Chemical Mfg Cor
B1195100ML |
100 mL | Amber Glass Bottle |
Each for $239.03
|
|
||||
18611912
|
Spectrum Chemical Mfg Cor
B11951LT |
1 L | Amber Glass Bottle |
Each for $454.30
|
|
||||
18611914
|
Spectrum Chemical Mfg Cor
B119525LTPV |
2.5 L | Poly-Coated Glass Bottle |
Each for $1,035.93
|
|
||||
18611915
|
Spectrum Chemical Mfg Cor
B119520LTBL |
20 L | Poly Drum |
Each for $4,007.90
|
|
||||
18611916
|
Spectrum Chemical Mfg Cor
B1195200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
107-92-6 |
Specifications
107-92-6 | |
98% | |
100 mL |
1 | |
Reagent | |
Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.