missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl Alcohol, B&J Brand™, for HPLC, GC and spectrophotometry, ≥99.9%, Honeywell Burdick & Jackson

$1290.53 - $1993.15

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6000092
View Documents
Honeywell-Burdick and Jackson
0241L
6 x 1 L Glass Bottle
Case of 6 Each for $1,290.53
Only null left
 
6000091
View Documents
Honeywell-Burdick and Jackson
0244
4 x 4 L Glass Bottle
Case of 4 Each for $1,993.15
Only null left
 
Description

Description

Specifications

Chemical Identifiers

71-36-3
74.123
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
CHEBI:28885
CCCCO
C4H10O
LRHPLDYGYMQRHN-UHFFFAOYSA-N
263
butan-1-ol

Specifications

71-36-3
100.0000%
117°C
37°C
C4H10O
0.03% max.
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
LRHPLDYGYMQRHN-UHFFFAOYSA-N
butan-1-ol
263
74.12
HPLC
0.5 AU max. at 225 nm (UV), 0.04 AU max. at 250 nm (UV), 0.01 AU max. at 275 nm (UV), 0.005 AU max. at 300 nm (UV), 0.005 AU max. at 400 nm (UV)
0.81 g/cm3
6 x 1 L
Glass Bottle
UN1120
969148
781% in Water at 20°C
CCCCO
74.123
CHEBI:28885
>99%
n-Butyl Alcohol, B&J Brand™, for HPLC, GC and spectrophotometry, ≥99.9%
Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Butanols

SDS
missing translation for 'documents'

missing translation for 'documents'