missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl Acetate ASTM D4615, Reagents

n-Butyl Acetate ASTM D4615, ASTM, for general laboratory use. Manufactured in ISO 9001 facility.

$82.07 - $390.10

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym Butyl Ethanoate, Acetic Acid Butyl Ester
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
01-337-381
View Documents
Labchem Inc
C21563004A
4 L Poly Bottle
Each for $390.10
Only null left
 
01-337-380
View Documents
Labchem Inc
C21563004C
4 L Glass Bottle
Each for $390.10
Only null left
 
01-337-379
View Documents
Labchem Inc
C2156300500C
500 mL Glass Bottle
Each for $82.07
Only null left
 
Description

Description

Specifications

Chemical Identifiers

123-86-4
116.16
Butyl Ethanoate, Acetic Acid Butyl Ester
CCCCOC(C)=O
C6H12O2
DKPFZGUDAPQIHT-UHFFFAOYSA-N
butyl acetate

Specifications

123-86-4
-78°C
126°C
Poly Bottle
4 L
Butyl Ethanoate, Acetic Acid Butyl Ester
CCCCOC(C)=O
116.16
n-Butyl Acetate ASTM D4615
126°C
Colorless
-78°C
C6H12O2
0.88
DKPFZGUDAPQIHT-UHFFFAOYSA-N
butyl acetate
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'