missing translation for 'onlineSavingsMsg'
Learn More

N-Bromophthalimide, 98+%

CAS: 2439-85-2 | C8H4BrNO2 | 226.029 g/mol

$59.66 - $556.07

Chemical Identifiers

CAS 2439-85-2
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.029
MDL Number MFCD00005888
InChI Key MARXMDRWROUXMD-UHFFFAOYSA-N
PubChem CID 75542
IUPAC Name 2-bromoisoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1160106
View Documents
Thermo Scientific Chemicals
A1160106
5 g
Each for $59.66
Only null left
 
AAA1160114
View Documents
Thermo Scientific Chemicals
A1160114
25 g
Each for $195.95
Only null left
 
AAA1160122
View Documents
Thermo Scientific Chemicals
A1160122
100 g
Each for $556.07
Only null left
 
Description

Description

Brominating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2439-85-2
226.029
MARXMDRWROUXMD-UHFFFAOYSA-N
2-bromoisoindole-1,3-dione
C8H4BrNO2
MFCD00005888
75542
C1=CC=C2C(=C1)C(=O)N(C2=O)Br

Specifications

2439-85-2
C8H4BrNO2
5 g
Light and moisture sensitive
MARXMDRWROUXMD-UHFFFAOYSA-N
2-bromoisoindole-1,3-dione
75542
≥98%
204°C to 208°C
MFCD00005888
131211
n-bromophthalimide, 2-bromoisoindoline-1,3-dione, 2-bromo-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 2-bromo, n-bromophtalimide, n-bromo phthalimide, acmc-1cjlt, 2-bromobenzo c azoline-1,3-dione, 1h-isoindole-1,3 2h-dione,2-bromo
C1=CC=C2C(=C1)C(=O)N(C2=O)Br
226.029
226.03
N-Bromophthalimide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 219-467-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only