missing translation for 'onlineSavingsMsg'
Learn More

N-Aminophthalimide, 94%

CAS: 1875-48-5 | C8H6N2O2 | 162.148 g/mol

Supplier:  Thermo Scientific Chemicals A1991409

 View more versions of this product

Catalog No. AAA1991409


Only null left
Explore available promotions

Description

Description

It is employed in the aziridination of chiral N-enoyl sultams. It is also used in the synthesis of n-phthalimidoaziridines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the aziridination of chiral N-enoyl sultams. It is also used in the synthesis of n-phthalimidoaziridines

Solubility
Solubility in hot methanol almost transparent.

Notes
Store away from oxidizing agents, strong bases and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications

Specifications

N-Aminophthalimide
∼195°C (decomposition)
MFCD00005895
383756
KSILMCDYDAKOJD-UHFFFAOYSA-N
2-aminoisoindole-1,3-dione
74645
94%
1875-48-5
C8H6N2O2
10 g
n-aminophthalimide, 2-aminoisoindoline-1,3-dione, 2-amino-1h-isoindole-1,3 2h-dione, phthalimide, n-amino, hydrazine, 1,1-phthaloyl, phthalhydrazide van, 1h-isoindole-1,3 2h-dione, 2-amino, n-aminophthalamide, phthalocyclohydrazide van, ccris 5349
C1=CC=C2C(=C1)C(=O)N(C2=O)N
162.148
162.15

Chemical Identifiers

1875-48-5
162.148
KSILMCDYDAKOJD-UHFFFAOYSA-N
74645
C1=CC=C2C(=C1)C(=O)N(C2=O)N
C8H6N2O2
MFCD00005895
n-aminophthalimide, 2-aminoisoindoline-1,3-dione, 2-amino-1h-isoindole-1,3 2h-dione, phthalimide, n-amino, hydrazine, 1,1-phthaloyl, phthalhydrazide van, 1h-isoindole-1,3 2h-dione, 2-amino, n-aminophthalamide, phthalocyclohydrazide van, ccris 5349
2-aminoisoindole-1,3-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 217-505-9

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only