missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N(alpha)-Boc-D-tryptophanol, 98%
$473.10 - $473.10
Chemical Identifiers
CAS | 158932-00-4 |
---|---|
Molecular Formula | C16H22N2O3 |
Molecular Weight (g/mol) | 290.36 |
MDL Number | MFCD00270221 |
InChI Key | JEFQUFUAEKORKL-GFCCVEGCSA-N |
Synonym | boc-d-tryptophanol, n-boc-d-tryptophanol, r-tert-butyl 1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, n alpha-boc-d-tryptophanol, boc-d-trp-ol, tert-butyl n-2r-1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, ambotzbal1034, n-t-boc-d-tryptophanol, tert-butyl r-1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, tert-butyl 2r-1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate |
PubChem CID | 20744818 |
SMILES | CC(C)(C)OC(=O)N[C@@H](CO)CC1=CNC2=CC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2776303
|
Thermo Scientific Chemicals
H2776303 |
1 g |
Each for $473.10
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
158932-00-4 | |
290.36 | |
JEFQUFUAEKORKL-GFCCVEGCSA-N | |
20744818 |
C16H22N2O3 | |
MFCD00270221 | |
boc-d-tryptophanol, n-boc-d-tryptophanol, r-tert-butyl 1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, n alpha-boc-d-tryptophanol, boc-d-trp-ol, tert-butyl n-2r-1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, ambotzbal1034, n-t-boc-d-tryptophanol, tert-butyl r-1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, tert-butyl 2r-1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate | |
CC(C)(C)OC(=O)N[C@@H](CO)CC1=CNC2=CC=CC=C12 |
Specifications
158932-00-4 | |
1 g | |
C16H22N2O3 | |
boc-d-tryptophanol, n-boc-d-tryptophanol, r-tert-butyl 1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, n alpha-boc-d-tryptophanol, boc-d-trp-ol, tert-butyl n-2r-1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, ambotzbal1034, n-t-boc-d-tryptophanol, tert-butyl r-1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, tert-butyl 2r-1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate | |
CC(C)(C)OC(=O)N[C@@H](CO)CC1=CNC2=CC=CC=C12 | |
290.36 | |
290.36 | |
N(alpha)-Boc-D-tryptophanol |
119°C to 121°C | |
+53° (c=2 in Acetic Acid) | |
MFCD00270221 | |
JEFQUFUAEKORKL-GFCCVEGCSA-N | |
tert-butyl N-[(2R)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate | |
20744818 | |
98% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only