missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-trans-(tert-butoxy)-L-proline 98.0+%, TCI America™
$61.51 - $257.24
Chemical Identifiers
CAS | 122996-47-8 |
---|---|
Molecular Formula | C24H27NO5 |
Molecular Weight (g/mol) | 409.482 |
MDL Number | MFCD00151930 |
InChI Key | WPBXBYOKQUEIDW-FXMQYSIJSA-N |
Synonym | fmoc-4-tert-butoxy-l-proline, 2s-4-tert-butoxy-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, fmoc-o-tert-butyl-l-hydroxyproline, ksc491o9t, n-alpha-9-fluorenylmethyloxycarbonyl-o-t-butyl-trans-l-hydroxyproline, 2s-1-9h-fluoren-9-ylmethoxycarbonyl-4-2-methylpropan-2-yl oxy pyrrolidine-2-carboxylic acid |
PubChem CID | 6916162 |
IUPAC Name | (2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-[(2-methylpropan-2-yl)oxy]pyrrolidine-2-carboxylic acid |
SMILES | CC(C)(C)OC1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O |
Chemical Identifiers
122996-47-8 | |
409.482 | |
WPBXBYOKQUEIDW-FXMQYSIJSA-N | |
6916162 | |
CC(C)(C)OC1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O |
C24H27NO5 | |
MFCD00151930 | |
fmoc-4-tert-butoxy-l-proline, 2s-4-tert-butoxy-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, fmoc-o-tert-butyl-l-hydroxyproline, ksc491o9t, n-alpha-9-fluorenylmethyloxycarbonyl-o-t-butyl-trans-l-hydroxyproline, 2s-1-9h-fluoren-9-ylmethoxycarbonyl-4-2-methylpropan-2-yl oxy pyrrolidine-2-carboxylic acid | |
(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-[(2-methylpropan-2-yl)oxy]pyrrolidine-2-carboxylic acid |
Specifications
122996-47-8 | |
White-Yellow | |
MFCD00151930 | |
fmoc-4-tert-butoxy-l-proline, 2s-4-tert-butoxy-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, fmoc-o-tert-butyl-l-hydroxyproline, ksc491o9t, n-alpha-9-fluorenylmethyloxycarbonyl-o-t-butyl-trans-l-hydroxyproline, 2s-1-9h-fluoren-9-ylmethoxycarbonyl-4-2-methylpropan-2-yl oxy pyrrolidine-2-carboxylic acid | |
CC(C)(C)OC1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O | |
409.482 | |
409.48 | |
Crystalline Powder |
62°C | |
C24H27NO5 | |
1 g | |
WPBXBYOKQUEIDW-FXMQYSIJSA-N | |
(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-[(2-methylpropan-2-yl)oxy]pyrrolidine-2-carboxylic acid | |
6916162 | |
≥98.0% (HPLC) | |
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-trans-(tert-butoxy)-L-proline |
Safety and Handling
TSCA : No