missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ MTS™ Itraconazole [ITC] 0.002-32 μg/mL
MIC Test Strips are a quantitative assay for determining the Minimum Inhibitory Concentration (MIC) of antimicrobial agents against microorganisms to indicate appropriate patient treatment and for identifying resistance patterns.
$315.23 - $315.23
Specifications
Antimicrobial Agents | Itraconazole |
---|---|
Concentration | 0.002 to 32 μg/mL |
Content And Storage | -20°C or 2°C to 8°C until the given expiry date |
Description | Liofilchem™ MTS™ Itraconazole |
Antibiotic Code | ITC |
Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity | ||||
22-777-968
![]() |
Liofilchem™
921481 |
10/Pk. | Individually Packed |
Pack of 10 Each for $315.23
|
|
||||
Description
Specifications
Itraconazole | |
-20°C or 2°C to 8°C until the given expiry date | |
ITC | |
Antifungal Susceptibility Testing, For determining the MIC of antimicrobial agents against microorganisms |
0.002 to 32 μg/mL | |
Liofilchem™ MTS™ Itraconazole | |
MIC Test Strip |
Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.