missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 50X HT Medium Supplement

Supplier:  MP Biomedicals™ 091680949

Catalog No. MP091680949


Only null left
Explore available promotions

Description

Description

  • 50X Concentrate
  • Hypoxanthine: 13.61mg/L
  • Thymidine: 3.876mg/L

Specifications

Specifications

0° to -20°C
Sterile
100 mL
HT Supplement
8.8 to 10.4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions