Learn More
Invitrogen™ Mouse IgM Isotype Control (11E10), FITC, eBioscience™
Mouse Monoclonal Isotype Control
Supplier: Invitrogen™ 11475280
Description
Description: The mouse IgM is useful as an isotype control immunoglobulin. The immunoglobulin was generated by immunization with lipopolysaccharide (LPS). The immunoglobulin is reactive with LPS. Applications Reported: This mouse IgM isotype control has been reported for use in immunocytochemistry, immunohistochemistry, and flow cytometric analysis. Applications Tested: This mouse IgM isotype control has been pre-titrated and tested by flow cytometric analysis of human peripheral blood leukocytes and can be used at the same concnetration as the experimental antibody. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgM is an immunoglobulin M antibody that is produced by B cells in response to an antigen. It is the first antibody produced during an immune response and is the largest immunoglobulin in the serum. Mouse IgM is composed of five heavy chains and five light chains, and it is involved in complement activation and agglutination of antigens. It plays a crucial role in the innate immune response and is important for neutralizing pathogens before the adaptive immune response is fully activated. Mouse IgM is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry.Specifications
Mouse IgM | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
25 μg | |
Isotype Control (Primary) | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Control, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry | |
11E10 | |
FITC | |
AGM1; AI326478; Igh6; Igh-6; Ighm; Igh-M; Igm; immunoglobulin heavy constant mu; MU; muH; VH | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Isotype Control | |
IgM |