missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Moroxydine Hydrochloride 98.0+%, TCI America™
$98.79 - $186.41
Chemical Identifiers
CAS | 3160-91-6 |
---|---|
Molecular Formula | C6H14ClN5O |
Molecular Weight (g/mol) | 207.662 |
MDL Number | MFCD00012798 |
InChI Key | FXYZDFSNBBOHTA-UHFFFAOYSA-N |
Synonym | 1-Morpholinobiguanide Hydrochloride |
PubChem CID | 76621 |
IUPAC Name | N-(diaminomethylidene)morpholine-4-carboximidamide;hydrochloride |
SMILES | C1COCCN1C(=N)N=C(N)N.Cl |
Chemical Identifiers
3160-91-6 | |
207.662 | |
FXYZDFSNBBOHTA-UHFFFAOYSA-N | |
76621 | |
C1COCCN1C(=N)N=C(N)N.Cl |
C6H14ClN5O | |
MFCD00012798 | |
1-Morpholinobiguanide Hydrochloride | |
N-(diaminomethylidene)morpholine-4-carboximidamide;hydrochloride |
Specifications
3160-91-6 | |
White | |
MFCD00012798 | |
1-Morpholinobiguanide Hydrochloride | |
C1COCCN1C(=N)N=C(N)N.Cl | |
207.662 | |
207.66 | |
Crystalline Powder |
202°C | |
C6H14ClN5O | |
5 g | |
FXYZDFSNBBOHTA-UHFFFAOYSA-N | |
N-(diaminomethylidene)morpholine-4-carboximidamide;hydrochloride | |
76621 | |
≥98.0% (HPLC,T) | |
Moroxydine Hydrochloride |
Safety and Handling
RTECSNumber : QD9815000
TSCA : No