missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieves, 5Å, Honeywell Fluka™
Beads, 8-12 mesh
$564.05 - $1750.78
Chemical Identifiers
CAS | 69912-79-4 |
---|---|
Molecular Formula | AlNaO6Si2 |
Molecular Weight (g/mol) | 202.14 |
MDL Number | MFCD00147627 |
InChI Key | URGAHOPLAPQHLN-UHFFFAOYSA-N |
PubChem CID | 19758701 |
SMILES | [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
69912-79-4 | |
202.14 | |
URGAHOPLAPQHLN-UHFFFAOYSA-N | |
[Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O |
AlNaO6Si2 | |
MFCD00147627 | |
19758701 |
Specifications
8 to 12mesh | |
5 Å | |
Beads | |
AlNaO6Si2 | |
MFCD00147627 | |
URGAHOPLAPQHLN-UHFFFAOYSA-N | |
Molecular sieves 5A | |
19758701 |
1 kg | |
Poly Bottle | |
69912-79-4 | |
Ca/nNa12-2n[(AlO2)12(SiO2)12] · xH2O | |
NONH for all modes of transport | |
[Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O | |
202.14 | |
Molecular Sieves, 5Å |