missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIP-1 delta Monoclonal Antibody (K70043_4C5), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555636
Description
Sequence of this protein is as follows: QFTNDAETEL MMSKLPLENP VVLNSFHFAA DCCTSYISQS IPCSLMKSYF ETSSECSKPG VIFLTKKGRQ VCAKPSGPGV QDCMKKLKPY SI
This gene is located in a cluster of similar genes in the same region of chromosome 17. These genes encode CC cytokines, which are secreted proteins characterized by two adjacent cysteines. The product of this gene is chemotactic for T cells and monocytes, and acts through C-C chemokine receptor type 1 (CCR1). The proprotein is further processed into numerous smaller functional peptides. Naturally-occurring readthrough transcripts occur from this gene into the downstream gene, CCL14 (chemokine (C-C motif) ligand 14).Specifications
MIP-1 delta | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q16663 | |
CCL15 | |
Recombinant human CCL15 (immunogen range is 22-113). | |
50 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Western Blot | |
K70043_4C5 | |
Unconjugated | |
CCL15 | |
C-C motif chemokine 15; C-C motif chemokine ligand 15; CCL15; CCL15(22-92); CCL15(25-92); CCL15(29-92); chemokine (C-C motif) ligand 15; chemokine CC-2; HCC-2; HCC-2Lkn-1; HMRP-2B; leukotactin 1; Leukotactin-1; LKN1; LKN-1; Macrophage inflammatory protein 5; MIP-1 delta; MIP-1D; MIP5; MIP-5; MIP-5MIP-1d; Mrp-2b; NCC3; NCC-3; NCC-3MIP-1D; NCC3MRP-2B; new CC chemokine 3; SCYA15; SCYA15member 15; SCYL3; small inducible cytokine subfamily A (Cys-Cys), member 15; small-inducible cytokine A15; SY15 | |
Mouse | |
Protein A | |
RUO | |
6359 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |