Learn More
Ted Pella Inc Microscope Slide Mailer, black DFS Item
Supplier:  Ted Pella Inc TPI22519
Disposable staining container is also useful for shipping, storing and handling of slides. Polypropylene container holds 5 3 x 1" slides in place so they will not touch. Positive lock and flip-top.
Catalog No. NC1255717
Only null left
Explore available promotions