missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ MicroGuard MP Microporous Shoe Cover with Non-skid Sole and Elastic Closure
Using state-of-the-art lamination technology, MicroGuard MP microporous shoe covers combine a soft, non-woven fabric with a microporous film to protect against spills and splashes.
$199.26 - $254.41
Specifications
Material | Multi-layer Barrier Laminate Fabric, Spunbond Polypropylene |
---|---|
Latex Free | Yes |
Color | White |
Disposable | Single-use |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17-184-068
|
International Enviroguard™
IEG8103 |
Large |
Case of 200 Each for $199.26
|
|
|||||
17-184-066
|
International Enviroguard™
8105 |
X-Large |
Case of 200 Each for $254.41
|
|
|||||
Description
- Shoe covers are designed with a non-skid sole that helps reduce the risk of slips or falls
- A tear-resistant liquid and particulate barrier helps safeguard from blood penetration, various industrial chemicals, along with dust, dirt, and grime
- Breathable fabric allows moisture to escape, so your feet stay cool and comfortable even as you sweat
- Strong elastic closure on top helps keep these durable shoe covers in place, so your shoes and feet are protected throughout your workday
- Environmentally friendly; can be recycled after use
- Made of lightweight, tear-resistant polypropylene
- Passes ASTM F1670 testing for blood penetration
Specifications
Multi-layer Barrier Laminate Fabric, Spunbond Polypropylene | |
White | |
Non-sterile | |
Serged |
Yes | |
Single-use | |
Elastic Top | |
Shoe Covers |