missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ MicroGuard MP 17 in. Microporous Boot Cover with Non-skid Sole and Elastic Closure
Using state-of-the-art lamination technology, MicroGuard MP microporous boot covers combine a soft, non-woven fabric with a microporous film that protects against splashes and spills.
Supplier: International Enviroguard™ 8106
Description
- Boot covers are designed with a non-skid sole to help reduce the risk of slips or falls
- A tear-resistant liquid and particulate barrier helps safeguard from blood penetration, various industrial chemicals, along with dust, dirt, and grime
- Breathable fabric allows moisture to escape, so your feet stay more cool and comfortable even as you sweat
- 17 in. length helps protect more of your lower legs from exposure to on-the-job contaminants
- Secure elastic closure helps keep boot covers in place throughout your workday
- Environmentally friendly; can be recycled after use
- Strong, elastic top for a secure fit
- Made of lightweight, tear-resistant polypropylene
- Passes ASTM F1670 testing for blood penetration
Specifications
X-Large | |
Yes | |
Single-use | |
Elastic Top with Ankle Ties | |
Serged | |
Boot Covers |
Multi-layer Barrier Laminate Fabric, Spunbond Polypropylene | |
White | |
Non-sterile | |
Non-skid | |
Fluid, Dry Particulates, Grime |