missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 Ts Plus Coveralls
Breathable, microporous laminate suit protects against liquid and particulate biological hazards including radioactive particulates. Previously known as MICROCHEM™ by AlphaTec 2000 STANDARD.
$431.99 - $577.87
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Material | Microporous Polyethylene Laminate (Non-woven) |
Color | White |
Disposable | Limited-use |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-500-822
|
Ansell
WH20T9211102 |
Small |
Case of 25 Each for $459.41
|
|
|||||
19-500-823
|
Ansell
WH20T9211103 |
Medium |
Case of 25 Each for $431.99
|
|
|||||
19-500-824
|
Ansell
WH20T9211104 |
Large |
Case of 25 Each for $431.99
|
|
|||||
19-500-825
|
Ansell
WH20T9211105 |
X-Large |
Case of 25 Each for $459.41
|
|
|||||
19-500-826
|
Ansell
WH20T9211106 |
2X-Large |
Case of 25 Each for $431.99
|
|
|||||
19-500-827
|
Ansell
WH20T9211107 |
3X-Large |
Case of 25 Each for $486.58
|
|
|||||
19-500-828
|
Ansell
WH20T9211108 |
4X-Large |
Case of 25 Each for $535.85
|
|
|||||
19-500-829
|
Ansell
WH20T9211109 |
5X-Large |
Case of 25 Each for $577.87
|
|
|||||
Description
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Anti-static, tested according to EN 1149-5 and AATTC 76
- Ultra Low Linting, reduced risk of contamination in critical areas
- Silicone free, critical in spray painting applications
- Elasticated hood, wrists, and ankles help to minimize the risk of linting and cross contamination
- Free of natural rubber latex
Specifications
General Purpose Coveralls | |
Microporous Polyethylene Laminate (Non-woven) | |
Limited-use | |
Elastic | |
Hooded | |
Front Zipper with Storm Flap | |
Non-sterile | |
Hooded | |
Yes | |
None | |
25/Cs. |
Unisex | |
White | |
Elastic | |
Thumb Loop | |
3-panel, Respirator Fit | |
Serged, Taped | |
Liquid, Particle, Bloodborne Pathogens | |
CE CATEGORY III (Type 4-B, Type 5-B, Type 6-B, EN 1073-2, EN 14126, DIN 32781, EN 1149-5) | |
WH20-T-92-111 | |
Life Sciences, Automotive, Metal Fabrication, Food Processing, Utilities, Oil and Gas, Machinery, Chemical |