Learn More
Mettler Toledo™ Rondolino 9-Place Sample Changer
Carousel-type titration stand to automize titrations with Mettler Toledo general titrators
Supplier: Mettler Toledo™ 51108500
Description
Can be loaded with up to nine sample beakers and one conditioning beaker in a unique position.
Plug it in and off you go
- Installation and operation could not be easier: simply connect it to the titrator and off you go
- No need to configure anything, no complicated settings, no time-wasting
- You can also select one of eight different conditioning and rinsing programs with a clearly marked rotating switch on the upper side of the instrument
- Requires less space than the typical sample changer—an area of 25 x 25cm is sufficient
- Handles series of up to nine samples
- Processes single samples more rapidly because beaker returned to the rack; after titration, electrodes are placed back carefully into conditioning beaker
- Reproducibility of results is improved because the measurement process is repeated exactly each time
- Cleans electrode quickly by dipping into conditioning beaker
- Suitable for most applications with a minimal carryover
Specifications
Sample Changer |