missing translation for 'onlineSavingsMsg'
Learn More

Metrizoic Acid 98.0+%, TCI America™

$57.79 - $109.98

Chemical Identifiers

CAS 1949-45-7
Molecular Formula C12H11I3N2O4
Molecular Weight (g/mol) 627.94
MDL Number MFCD00867964
InChI Key GGGDNPWHMNJRFN-UHFFFAOYSA-N
Synonym 3-Acetamido-5-(N-methylacetamido)-2,4,6-triiodobenzoic Acid
PubChem CID 2528
ChEBI CHEBI:34847
IUPAC Name 3-acetamido-2,4,6-triiodo-5-(N-methylacetamido)benzoic acid
SMILES CN(C(C)=O)C1=C(I)C(NC(C)=O)=C(I)C(C(O)=O)=C1I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M13635G
View Documents
TCI America
M13635G
5 g
Each for $57.79
Only null left
 
M136325G
View Documents
TCI America
M136325G
25 g
Each for $109.98
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1949-45-7
627.94
GGGDNPWHMNJRFN-UHFFFAOYSA-N
2528
3-acetamido-2,4,6-triiodo-5-(N-methylacetamido)benzoic acid
C12H11I3N2O4
MFCD00867964
3-Acetamido-5-(N-methylacetamido)-2,4,6-triiodobenzoic Acid
CHEBI:34847
CN(C(C)=O)C1=C(I)C(NC(C)=O)=C(I)C(C(O)=O)=C1I

Specifications

1949-45-7
C12H11I3N2O4
5 g
GGGDNPWHMNJRFN-UHFFFAOYSA-N
3-acetamido-2,4,6-triiodo-5-(N-methylacetamido)benzoic acid
2528
627.94
Crystalline Powder
White
MFCD00867964
3-Acetamido-5-(N-methylacetamido)-2,4,6-triiodobenzoic Acid
CN(C(C)=O)C1=C(I)C(NC(C)=O)=C(I)C(C(O)=O)=C1I
627.94
CHEBI:34847
≥98.0% (T,HPLC)
Metrizoic Acid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : DG0887300

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'