missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metoprolol Tartrate 98.0+%, TCI America™
Supplier: TCI America M255525G
Specifications
Metoprolol Tartrate | |
123°C | |
C34H56N2O12 | |
25 g | |
YGULWPYYGQCFMP-NPHUUBOGSA-N | |
(2R,3S)-2,3-dihydroxybutanedioic acid;1-[4-(2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol | |
11957594 | |
≥98.0% (HPLC,T) |
56392-17-7 | |
White | |
MFCD00056257 | |
y-metoprolol +-tartrate, ?-metoprolol +-tartrate, --tartaric acid; bis metoprolol, 1-isopropylamino-3-p-beta-methoxyethyl phenoxy-2-propanol +-tartrate salt, 1-isopropylamino-3-4-2-methoxyethyl phenoxy propan-2-ol 2,3-dihydroxysuccinate 2 | |
CC(C)NCC(COC1=CC=C(C=C1)CCOC)O.CC(C)NCC(COC1=CC=C(C=C1)CCOC)O.C(C(C(=O)O)O)(C(=O)O)O | |
684.824 | |
342.41 | |
Crystalline Powder |
Chemical Identifiers
56392-17-7 | |
684.824 | |
YGULWPYYGQCFMP-NPHUUBOGSA-N | |
11957594 | |
CC(C)NCC(COC1=CC=C(C=C1)CCOC)O.CC(C)NCC(COC1=CC=C(C=C1)CCOC)O.C(C(C(=O)O)O)(C(=O)O)O |
C34H56N2O12 | |
MFCD00056257 | |
y-metoprolol +-tartrate, ?-metoprolol +-tartrate, --tartaric acid; bis metoprolol, 1-isopropylamino-3-p-beta-methoxyethyl phenoxy-2-propanol +-tartrate salt, 1-isopropylamino-3-4-2-methoxyethyl phenoxy propan-2-ol 2,3-dihydroxysuccinate 2 | |
(2R,3S)-2,3-dihydroxybutanedioic acid;1-[4-(2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol |
Safety and Handling
RTECSNumber : UB7450100
TSCA : No