missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metoclopramide hydrochloride, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 7232-21-5 |
---|---|
Molecular Formula | C14H23Cl2N3O2 |
Molecular Weight (g/mol) | 336.26 |
InChI Key | RVFUNJWWXKCWNS-UHFFFAOYSA-N |
IUPAC Name | 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrochloride |
SMILES | Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC461820050
|
Thermo Scientific Chemicals
461820050 |
5 g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC461820250
|
Thermo Scientific Chemicals
461820250 |
25 g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7232-21-5 | |
336.26 | |
4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrochloride |
C14H23Cl2N3O2 | |
RVFUNJWWXKCWNS-UHFFFAOYSA-N | |
Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC |
Specifications
7232-21-5 | |
C14H23Cl2N3O2 | |
Solubility aq. soln.: soluble. Other solubilities: soluble in methanol | |
Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC | |
336.26 | |
Metoclopramide hydrochloride |
98% | |
5 g | |
RVFUNJWWXKCWNS-UHFFFAOYSA-N | |
4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrochloride | |
336.26 |
Safety and Handling
GHS H Statement Harmful if swallowed.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
EINECSNumber : 230-634-5
RUO â Research Use Only