missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylsulfoxide, 99.9% min., OmniSolv, MilliporeSigma™
$384.51 - $1012.00
Chemical Identifiers
CAS | 67-68-5 |
---|---|
Molecular Formula | C2H6OS |
Molecular Weight (g/mol) | 78.13 |
MDL Number | MFCD00002089 |
InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
Synonym | dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide |
PubChem CID | 679 |
ChEBI | CHEBI:28262 |
IUPAC Name | methanesulfinylmethane |
SMILES | CS(C)=O |
Chemical Identifiers
67-68-5 | |
78.13 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
679 | |
methanesulfinylmethane |
C2H6OS | |
MFCD00002089 | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CHEBI:28262 | |
CS(C)=O |
Specifications
18.4°C | |
Colorless | |
189°C | |
214.85°C (418.7°F) | |
Liquid | |
C2H6OS | |
MFCD00002089 | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CS(C)=O | |
78.13 | |
CHEBI:28262 | |
Methylsulfoxide |
67-68-5 | |
1.10g/cm3 at 20°C | |
1 L | |
Poly-coated Glass Bottle | |
99.9% min. | |
(CH3)2SO | |
0.053hPa (20°C) | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
methanesulfinylmethane | |
679 | |
78.1 |
Safety and Handling
Recommended Storage : Store at +15°C to +25°C