missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylene Chloride, NF, 99%, Spectrum™ Chemical
M1266, 75-09-2, CH2Cl2
Supplier: Spectrum Chemical Mfg Cor M1266500MLGLCS6
Description
Spectrum™ Chemical Methylene Chloride, NF is used as a carrier for pharmaceutical tablet coatings. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
75-09-2 | |
6 x 500 mL | |
CH2Cl2 | |
UN1593 | |
ClCCl | |
84.93 |
100% | |
Amber Glass Bottle | |
MFCD00000881 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
99% |
Chemical Identifiers
75-09-2 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
ClCCl |
CH2Cl2 | |
MFCD00000881 | |
dichloromethane |