missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylene Chloride, FCC, 99%, Spectrum™ Chemical
M1172, 75-09-2, CH2Cl2
Supplier: Spectrum Chemical Mfg Cor M11724LT
Description
Spectrum™ Chemical Methylene Chloride, FCC is used to decaffeinate coffee and tea as well as to prepare extracts of hops and other flavorings. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
75-09-2 | |
100% | |
Amber Glass Bottle | |
CH2Cl2 | |
UN1593 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
99% |
10 mg/kg | |
4 L | |
1.318-1.323 | |
MFCD00000881 | |
0.02% | |
ClCCl | |
84.93 | |
FCC |
Chemical Identifiers
75-09-2 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
ClCCl |
CH2Cl2 | |
MFCD00000881 | |
dichloromethane |