missing translation for 'onlineSavingsMsg'
Learn More

Methylbenzethonium Chloride, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor M1244100GMBL

 View more versions of this product

Catalog No. 18611759

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

25155-18-4
Poly Bottle
0.98
Crystalline Powder or Lumps
1
100 g
Reagent

Chemical Identifiers

25155-18-4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.