missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfoxide, 99.9+%, ACS reagent, Thermo Scientific Chemicals

$242.69 - $515.24

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC414885000
SDS View Documents
Thermo Scientific Chemicals
414885000
500 mL EcoSafPak™ Plastic Bottle
Each for $242.69
Case of 6 Each for $1,456.14
Only null left
 
AC414880010
SDS View Documents
Thermo Scientific Chemicals
414880010
1 L Plastic Bottle
Each for $273.60
Only null left
 
AC414880025
SDS View Documents
Thermo Scientific Chemicals
414880025
2.5 L Plastic Bottle
Each for $515.24
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67-68-5
78.13
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
methylsulfinylmethane
C2H6OS
MFCD00002089
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CHEBI:28262
CS(C)=O

Specifications

18.4°C
Colorless
100.0
1.1000g/mL
500 mL
EcoSafPak™ Plastic Bottle
1.4770 to 1.4800
99.9+%
0.001 meq/g max.
(CH3)2SO
Vapor Pressure: 0.55mbar at 20°C
0.1% max. (Coulometric)
01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methylsulfinylmethane
679
78.13
ACS Reagent
67-68-5
99.9
>190°C
189.0°C
87°C
Liquid
0.01% max.
1.1
C2H6OS
MFCD00002089
1.987 mPa.s (25°C)
01, IV, 1277
15, 3285
Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons
CS(C)=O
78.13
CHEBI:28262
≥99.9%
Methyl sulfoxide
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

EINECSNumber : 200-664-3

RTECSNumber : PV6210000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only