missing translation for 'onlineSavingsMsg'
Learn More

Methyl Iodide, 99.5%, Spectrum™ Chemical

$208.30 - $34626.50

Chemical Identifiers

CAS 74-88-4
Molecular Formula CH3I
Molecular Weight (g/mol) 141.94
InChI Key INQOMBQAUSQDDS-UHFFFAOYSA-N
IUPAC Name iodomethane
SMILES CI
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611701
View Documents
Spectrum Chemical Mfg Cor
M127025GM
25 g Amber Glass Bottle
Each for $208.30
Only null left
 
18611700
View Documents
Spectrum Chemical Mfg Cor
M1270100GM
100 g Amber Glass Bottle
Each for $477.23
Only null left
 
18611702
View Documents
Spectrum Chemical Mfg Cor
M1270500GM
500 g Amber Glass Bottle
Each for $1,526.77
Only null left
 
18611703
View Documents
Spectrum Chemical Mfg Cor
M127060KGBL
60 kg Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

74-88-4
141.94
iodomethane
CH3I
INQOMBQAUSQDDS-UHFFFAOYSA-N
CI

Specifications

74-88-4
Amber Glass Bottle
25 g
INQOMBQAUSQDDS-UHFFFAOYSA-N
iodomethane
0.995
1
CH3I
0.0001
CI
141.94
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.